Tisk…

3,3,4,4,5,5-Hexafluoropentan-1-ol

Dodavatel: Apollo Scientific

PC14286-100MG PC14286-25MG
APOSPC14286-100MGEA 29200 CZK
APOSPC14286-100MG APOSPC14286-25MG
3,3,4,4,5,5-Hexafluoropentan-1-ol
3,3,4,4,5,5-Hexafluoropentan-1-ol
Vzorec: C₅H₆F₆O
MW: 196,091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31800421
CAS číslo: 1639207-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR