Tisk…

4-Chloro-2-fluorobenzyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α,4-Dichloro-2-fluorotoluene, 4-Chloro-1-(chloromethyl)-2-fluorobenzene

PC1241-1G PC1241-25G PC1241-5G
APOSPC1241-1GEA 622 CZK
APOSPC1241-1G APOSPC1241-25G APOSPC1241-5G
4-Chloro-2-fluorobenzyl chloride 95%
4-Chloro-2-fluorobenzyl chloride
Vzorec: C₇H₅Cl₂F
MW: 179,019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412453
CAS číslo: 87417-71-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR