Tisk…

3-Iodo-1,1,1-trifluoroacetone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Iodo-1,1,1-trifluoropropan-2-one, 3-Iodo-2-oxo-1,1,1-trifluoropropane

PC1233-1G PC1233-5G
APOSPC1233-1GEA 5990 CZK
APOSPC1233-1G APOSPC1233-5G
3-Iodo-1,1,1-trifluoroacetone
3-Iodo-1,1,1-trifluoroacetone
Vzorec: C₃H₂F₃IO
MW: 237,947 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09998066
CAS číslo: 142921-30-0
UN: 2924
ADR: 3+8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR