Tisk…
N,N-Dimethyl-1-[4-(trifluoromethyl)phenyl]ethane-1,2-diamine dihydrochloride
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: {2-Amino-1-[4-(trifluoromethyl)phenyl]ethyl}dimethylamine dihydrochloride, 4-[2-Amino-1-(dimethylamino)ethyl]benzotrifluoride dihydrochloride
PC1157-100MG PC1157-500MG PC1157-250MG
APOSPC1157-100MGEA 4960 CZK
APOSPC1157-100MG APOSPC1157-500MG APOSPC1157-250MG
N,N-Dimethyl-1-[4-(trifluoromethyl)phenyl]ethane-1,2-diamine dihydrochloride
N,N-Dimethyl-1-[4-(trifluoromethyl)phenyl]ethane-1,2-diamine dihydrochloride
Vzorec: C₁₁H₁₅F₃N₂·2ClH
MW: 305.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840183
CAS číslo: 1189496-82-9
UN: 3261
ADR: 8 III