Tisk…

N,N-Dimethyl-2-(trifluoromethoxy)benzylamine

Dodavatel: Apollo Scientific
Varování

Synonyms: N,N-Dimethyl-1-[2-(trifluoromethoxy)phenyl]methylamine

PC11281-25G PC11281-5G
APOSPC11281-25GEA 23610 CZK
APOSPC11281-25G APOSPC11281-5G
N,N-Dimethyl-2-(trifluoromethoxy)benzylamine
N,N-Dimethyl-2-(trifluoromethoxy)benzylamine
Vzorec: C₁₀H₁₂F₃NO
MW: 219,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368741
CAS číslo: 886762-84-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR