Tisk…

1-[3-(Trifluoromethoxy)benzyl]piperazine

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-{[3-(Trifluoromethoxy)phenyl]methyl}piperazine

PC11280-1G PC11280-25G PC11280-5G
APOSPC11280-1GEA 1450 CZK
APOSPC11280-1G APOSPC11280-25G APOSPC11280-5G
1-[3-(Trifluoromethoxy)benzyl]piperazine
1-[3-(Trifluoromethoxy)benzyl]piperazine
Vzorec: C₁₂H₁₅F₃N₂O
MW: 260,256 g/mol
Bod varu: 82 °C (0,03 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07368740
CAS číslo: 868054-25-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR