4-Chloro-5-(4-fluorophenyl)thieno[2,3-d]pyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
PC11264-1G
PC11264-500MG
APOSPC11264-1GEA
6980
CZK
APOSPC11264-1G
APOSPC11264-500MG
4-Chloro-5-(4-fluorophenyl)thieno[2,3-d]pyrimidine
4-Chloro-5-(4-fluorophenyl)thieno[2,3-d]pyrimidine
Vzorec:
C₁₂H₆ClFN₂S Storage Temperature: Ambient |
MDL Number:
MFCD00297841 CAS číslo: 384351-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...