Tisk…

5,7-Difluorobenzo[b]thiophene-2-carboxylic acid

Dodavatel: Apollo Scientific

PC110115-1G PC110115-250MG
APOSPC110115-1GEA 11380 CZK
APOSPC110115-1G APOSPC110115-250MG
5,7-Difluorobenzo[b]thiophene-2-carboxylic acid
5,7-Difluorobenzo[b]thiophene-2-carboxylic acid
Vzorec: C₉H₄F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD13180848
CAS číslo: 550998-68-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR