Ethyl 6-bromo-8-fluoro-4-hydroxyquinoline-3-carboxylate
Dodavatel: Apollo Scientific
PC110037-1G
PC110037-250MG
APOSPC110037-1GEA
4770
CZK
APOSPC110037-1G
APOSPC110037-250MG
Ethyl 6-bromo-8-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 6-bromo-8-fluoro-4-hydroxyquinoline-3-carboxylate
Vzorec:
C₁₂H₉BrFNO₃ MW: 314,107 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25572662 CAS číslo: 228728-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...