Tisk…

8-Chloro-7-(trifluoromethyl)quinolin-4-ol

Dodavatel: Apollo Scientific
Varování

PC110013-1G PC110013-250MG
APOSPC110013-1GEA 6850 CZK
APOSPC110013-1G APOSPC110013-250MG
8-Chloro-7-(trifluoromethyl)quinolin-4-ol
8-Chloro-7-(trifluoromethyl)quinolin-4-ol
Vzorec: C₁₀H₅ClF₃NO
MW: 247,601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200325
CAS číslo: 2407339-49-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR