Tisk…

Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate 95%

Dodavatel: Apollo Scientific
Varování

PC10854-1G PC10854-250MG
APOSPC10854-1GEA 17760 CZK
APOSPC10854-1G APOSPC10854-250MG
Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate 95%
Methyl 5-fluoro-2-(trifluoromethyl)isonicotinate
Vzorec: C₈H₅F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD28799126
CAS číslo: 1803834-94-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR