Tisk…

2-(4-Amino-3-fluoro-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%

Dodavatel: Apollo Scientific
Varování

PC10737-1G PC10737-250MG
APOSPC10737-1GEA 1660 CZK
APOSPC10737-1G APOSPC10737-250MG
2-(4-Amino-3-fluoro-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(4-Amino-3-fluoro-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Vzorec: C₉H₅F₇N₂O₃
MW: 322,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066747
CAS číslo: 2366994-16-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR