Tisk…

2-(4-Bromo-3-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%

Dodavatel: Apollo Scientific
Varování

PC10736-1G PC10736-250MG PC10736-5G
APOSPC10736-1GEA 1580 CZK
APOSPC10736-1G APOSPC10736-250MG APOSPC10736-5G
2-(4-Bromo-3-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(4-Bromo-3-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Vzorec: C₉H₄BrF₇O
MW: 341,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066741
CAS číslo: 1917307-13-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR