Tisk…

4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-{[2-(Trifluoromethyl)quinolin-4-yl]thio}ethylamine

PC1070V-1G PC1070V-10G
APOSPC1070V-1GEA 1120 CZK
APOSPC1070V-1G APOSPC1070V-10G
4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline 97%
4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline
Vzorec: C₁₂H₁₁F₃N₂S
MW: 272.289 g/mol
Bod tání: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00174223
CAS číslo: 175203-50-6
UN: 3259
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR