Tisk…

[(Chlorodifluoromethyl)sulphonyl]benzene 95%

Dodavatel: Apollo Scientific

PC10693-100MG PC10693-1G PC10693-250MG
APOSPC10693-100MGEA 1080 CZK
APOSPC10693-100MG APOSPC10693-1G APOSPC10693-250MG
[(Chlorodifluoromethyl)sulphonyl]benzene 95%
[(Chlorodifluoromethyl)sulphonyl]benzene
Vzorec: C₇H₅ClF₂O₂S
Bod tání: 31…34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18253567
CAS číslo: 930836-30-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR