(Chlorodifluoromethyl)trimethylsilane 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC10691-100MG
PC10691-1G
PC10691-250MG
APOSPC10691-100MGEA
7480
CZK
APOSPC10691-100MG
APOSPC10691-1G
APOSPC10691-250MG
(Chlorodifluoromethyl)trimethylsilane 97%
(Chlorodifluoromethyl)trimethylsilane
Vzorec:
C₄H₉ClF₂Si MW: 158,65 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20486786 CAS číslo: 115262-00-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...