Tisk…

1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Bromophenyl)-1-(trifluoromethyl)cyclopropane, 1-Bromo-4-(1-(trifluoromethyl)cyclopropyl)benzene

PC10675-1G PC10675-5G
APOSPC10675-1GEA 5000 CZK
APOSPC10675-1G APOSPC10675-5G
1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene
1-Bromo-4-[1-(trifluoromethyl)cycloprop-1-yl]benzene
Vzorec: C₁₀H₈BrF₃
MW: 265,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817615
CAS číslo: 1227160-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR