3-Bromo-4-(trifluoromethyl)benzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC10671-10G
PC10671-1G
PC10671-5G
PC10671-25G
APOSPC10671-10GEA
15410
CZK
APOSPC10671-10G
APOSPC10671-1G
APOSPC10671-5G
APOSPC10671-25G
3-Bromo-4-(trifluoromethyl)benzaldehyde 98%
3-Bromo-4-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₄BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD13185384 CAS číslo: 372120-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...