Tisk…

5-[(Trifluoromethyl)sulphonyl]-1H-benzimidazole 95%

Dodavatel: Apollo Scientific
Varování

PC10660-1G PC10660-250MG
APOSPC10660-1GEA 5560 CZK
APOSPC10660-1G APOSPC10660-250MG
5-[(Trifluoromethyl)sulphonyl]-1H-benzimidazole 95%
5-[(Trifluoromethyl)sulphonyl]-1H-benzimidazole
Vzorec: C₈H₅F₃N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD22628760
CAS číslo: 1417341-55-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR