Tisk…

Perfluoro-3-methoxypropanoic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC10622-250MG PC10622-5G
APOSPC10622-250MGEA 4500 CZK
APOSPC10622-250MG APOSPC10622-5G
Perfluoro-3-methoxypropanoic acid 98%
Perfluoro-3-methoxypropanoic acid
Vzorec: C₄HF₇O₃
Bod varu: 126 °C (740 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00507228
CAS číslo: 377-73-1
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR