4-Methyl-2-(trifluoromethyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC10463-1G
PC10463-250MG
APOSPC10463-1GEA
20470
CZK
APOSPC10463-1G
APOSPC10463-250MG
4-Methyl-2-(trifluoromethyl)benzaldehyde
4-Methyl-2-(trifluoromethyl)benzaldehyde
Vzorec:
C₉H₇F₃O Storage Temperature: Ambient |
MDL Number:
MFCD18399857 CAS číslo: 1261622-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...