Tisk…

4-Bromo-3-fluorobenzene-1,2-diamine 98%

Dodavatel: Apollo Scientific
Varování

PC10402-1G PC10402-250MG PC10402-5G
APOSPC10402-1GEA 1240 CZK
APOSPC10402-1G APOSPC10402-250MG APOSPC10402-5G
4-Bromo-3-fluorobenzene-1,2-diamine 98%
4-Bromo-3-fluorobenzene-1,2-diamine
Vzorec: C₆H₆BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD07368683
CAS číslo: 886762-86-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR