Bis(1H,1H-heptafluorobutyl)amine
Dodavatel: Apollo Scientific
|
Varování
|
PC1024-1G
PC1024-5G
APOSPC1024-1GEA
6580
CZK
APOSPC1024-1G
APOSPC1024-5G
Bis(1H,1H-heptafluorobutyl)amine
Bis(1H,1H-heptafluorobutyl)amine
Vzorec:
C₈H₅F₁₄N MW: 381,11 g/mol Bod varu: 138…138,5 °C (735 mm Hg) Hustota: 1,614 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD07784276 CAS číslo: 356-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...