Perfluoroheptanenitrile
Dodavatel: Apollo Scientific
PC10009-1G
PC10009-5G
APOSPC10009-1GEA
3870
CZK
APOSPC10009-1G
APOSPC10009-5G
Perfluoroheptanenitrile
Perfluoroheptanenitrile
Vzorec:
C₇F₁₃N Bod varu: 82…83 °C Hustota: 1,6274 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03424480 CAS číslo: 62038-08-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...