Tisk…

Fluoroiodoacetic acid

Dodavatel: Apollo Scientific
Nebezpečí

PC0992-1G PC0992-5G
APOSPC0992-1GEA 5560 CZK
APOSPC0992-1G APOSPC0992-5G
Fluoroiodoacetic acid
Fluoroiodoacetic acid
Vzorec: C₂H₂FIO₂
MW: 203.939 g/mol
Bod tání: 79.5 °C
Bod vzplanutí: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784241
CAS číslo: 471-45-4
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR