4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC0956-1G
PC0956-250MG
PC0956-5G
APOSPC0956-1GEA
4570
CZK
APOSPC0956-1G
APOSPC0956-250MG
APOSPC0956-5G
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Vzorec:
C₁₂H₇F₃O₂S Bod tání: 130…132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00113429 CAS číslo: 208108-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...