Tisk…

2-[4-(Trifluoromethyl)phenoxy]acetohydrazide

Dodavatel: Apollo Scientific
Varování

Synonyms: [4-(Trifluoromethyl)phenoxy]acetic acid hydrazide, 4-(2-Hydrazino-2-oxoethoxy)benzotrifluoride

PC0933-1G PC0933-250MG
APOSPC0933-1GEA 3810 CZK
APOSPC0933-1G APOSPC0933-250MG
2-[4-(Trifluoromethyl)phenoxy]acetohydrazide
2-[4-(Trifluoromethyl)phenoxy]acetohydrazide
Vzorec: C₉H₉F₃N₂O₂
MW: 234,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035657
CAS číslo: 860649-71-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR