Tisk…

1-(Chlorodifluoromethyl)-4-(dichlorofluoromethyl)benzene 97%

Dodavatel: Apollo Scientific

PC0920-25G PC0920-5G PC0920-1G
APOSPC0920-25GEA 14090 CZK
APOSPC0920-25G APOSPC0920-5G APOSPC0920-1G
1-(Chlorodifluoromethyl)-4-(dichlorofluoromethyl)benzene 97%
1-(Chlorodifluoromethyl)-4-(dichlorofluoromethyl)benzene
Vzorec: C₈H₄Cl₃F₃
Bod varu: 88…89 °C (8 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07368668
CAS číslo: 13947-95-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR