2-Bromo-1,2,2-trifluoroethyl trifluoromethyl ether 97%
Dodavatel: Apollo Scientific
PC0833-1G
PC0833-5G
PC0833-25G
APOSPC0833-1GEA
2900
CZK
APOSPC0833-1G
APOSPC0833-5G
APOSPC0833-25G
2-Bromo-1,2,2-trifluoroethyl trifluoromethyl ether 97%
2-Bromo-1,2,2-trifluoroethyl trifluoromethyl ether
Vzorec:
C₃HBrF₆O MW: 246,934 g/mol Bod varu: 37…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368667 CAS číslo: 2356-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...