N-Trifluoroacetyl-L-cysteine methyl ester 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC0770-1G
PC0770-250MG
APOSPC0770-1GEA
29830
CZK
APOSPC0770-1G
APOSPC0770-250MG
N-Trifluoroacetyl-L-cysteine methyl ester 95%
N-Trifluoroacetyl-L-cysteine methyl ester
Vzorec:
C₆H₈F₃NO₃S MW: 231,193 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00055581 CAS číslo: 1577-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...