1,2,2,2-Tetrafluoroethyl methyl ether 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
,2,2,2-Tetrafluoro-1-methoxyethane, 1,1,1,2-Tetrafluoro-3-oxabutane
PC0764-25G
PC0764-5G
APOSPC0764-25GEA
35380
CZK
APOSPC0764-25G
APOSPC0764-5G
1,2,2,2-Tetrafluoroethyl methyl ether 99%
1,2,2,2-Tetrafluoroethyl methyl ether
Vzorec:
C₃H₄F₄O MW: 132.057 g/mol Bod varu: 38…39 °C (768 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07366529 CAS číslo: 50285-05-7 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...