Tisk…

3,5-Bis(trifluoromethyl)-1-phenylpyrazole

Dodavatel: Apollo Scientific
Varování

PC0625-1G PC0625-5G PC0625-250MG
APOSPC0625-1GEA 3510 CZK
APOSPC0625-1G APOSPC0625-5G APOSPC0625-250MG
3,5-Bis(trifluoromethyl)-1-phenylpyrazole
3,5-Bis(trifluoromethyl)-1-phenylpyrazole
Vzorec: C₁₁H₆F₆N₂
MW: 280,169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153671
CAS číslo: 140647-19-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR