3,3,3-Trifluoro-D-lactic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(2R)-2-Hydroxy-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-hydroxypropanoic acid
PC0618-1G
PC0618-250MG
APOSPC0618-1GEA
17960
CZK
APOSPC0618-1G
APOSPC0618-250MG
3,3,3-Trifluoro-D-lactic acid 97%
3,3,3-Trifluoro-D-lactic acid
Vzorec:
C₃H₃F₃O₃ MW: 144,049 g/mol Bod tání: 66…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093016 CAS číslo: 121250-04-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...