Tisk…

(S)-1-[3-(Trifluoromethyl)phenyl]ethanol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-1-(3-(Trifluoromethyl)phenyl)ethanol

PC0589-250MG PC0589-1G
APOSPC0589-250MGEA 5200 CZK
APOSPC0589-250MG APOSPC0589-1G
(S)-1-[3-(Trifluoromethyl)phenyl]ethanol 97%
(S)-1-[3-(Trifluoromethyl)phenyl]ethanol
Vzorec: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093004
CAS číslo: 96789-80-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR