Tisk…

1H-Nonafluorobutane 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,1,1,2,2,3,3,4,4-Nonafluorobutane

PC0548-10G PC0548-25G
APOSPC0548-10GEA 10750 CZK
APOSPC0548-10G APOSPC0548-25G
1H-Nonafluorobutane 99%
1H-Nonafluorobutane
Vzorec: C₄HF₉
MW: 220,036 g/mol
Bod varu: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00272355
CAS číslo: 375-17-7
UN: 3163
ADR: 2.2,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR