Methyl 3-methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-methyl-5-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylate, 4-[4-(Methoxycarbonyl)-3-methylisoxazol-5-yl]benzotrifluoride
PC0495-500MG
PC0495-1G
PC0495-5G
APOSPC0495-500MGEA
0
CZK
APOSPC0495-500MG
APOSPC0495-1G
APOSPC0495-5G
Methyl 3-methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylate
Methyl 3-methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylate
Vzorec:
C₁₃H₁₀F₃NO₃ MW: 285,219 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00100442 CAS číslo: 175276-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...