1-Chloro-8-iodoperfluorooctane 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC0473-25G
PC0473-5G
APOSPC0473-25GEA
7810
CZK
APOSPC0473-25G
APOSPC0473-5G
1-Chloro-8-iodoperfluorooctane 98%
1-Chloro-8-iodoperfluorooctane
Vzorec:
C₈ClF₁₆I MW: 562,418 g/mol Bod varu: 74…78 °C (4 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD01320707 CAS číslo: 16486-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...