4-Methyl-3-(trifluoromethyl)benzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-(Chlorocarbonyl)-2-methylbenzotrifluoride
PC0469-10G
PC0469-1G
PC0469-5G
APOSPC0469-10GEA
9430
CZK
APOSPC0469-10G
APOSPC0469-1G
APOSPC0469-5G
4-Methyl-3-(trifluoromethyl)benzoyl chloride
4-Methyl-3-(trifluoromethyl)benzoyl chloride
Vzorec:
C₉H₆ClF₃O MW: 222,592 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01631602 CAS číslo: 261952-11-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...