3-Methyl-5-(trifluoromethyl)benzamide
Dodavatel: Apollo Scientific
PC0442-10G
PC0442-5G
APOSPC0442-10GEA
14120
CZK
APOSPC0442-10G
APOSPC0442-5G
3-Methyl-5-(trifluoromethyl)benzamide
3-Methyl-5-(trifluoromethyl)benzamide
Vzorec:
C₉H₈F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD01631497 CAS číslo: 261951-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...