2-Methyl-4-(trifluoromethoxy)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethoxy)-o-tolunitrile, 2-Cyano-5-(trifluoromethoxy)toluene
PC0418-1G
PC0418-25G
PC0418-5G
APOSPC0418-1GEA
3770
CZK
APOSPC0418-1G
APOSPC0418-25G
APOSPC0418-5G
2-Methyl-4-(trifluoromethoxy)benzonitrile 98%
2-Methyl-4-(trifluoromethoxy)benzonitrile
Vzorec:
C₉H₆F₃NO MW: 201,145 g/mol Bod varu: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631543 CAS číslo: 261951-92-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...