Tisk…

α-(Trifluoromethyl)styrene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: [1-(Trifluoromethyl)vinyl]benzene, (3,3,3-Trifluoroprop-1-en-2-yl)benzene, Alpha-(trifluoromethyl)styrene

PC0401-1G
APOSPC0401-1GEA 7110 CZK
APOSPC0401-1G
α-(Trifluoromethyl)styrene 97%
α-(Trifluoromethyl)styrene
Vzorec: C₉H₇F₃
MW: 172,147 g/mol
Bod varu: 148…148,5 °C
Hustota: 1,167 g/cm³ (25 °C)
Bod vzplanutí: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081197
CAS číslo: 384-64-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR