Bis(fluoromethyl)ether
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0341-5G
PC0341-1G
APOSPC0341-5GEA
10980
CZK
APOSPC0341-5G
APOSPC0341-1G
Bis(fluoromethyl)ether
Bis(fluoromethyl)ether
Useful synthetic intermediate; call for data sheet.
Vzorec:
C₂H₄F₂O MW: 82,049 g/mol Bod varu: 34 °C Bod tání: –70 °C Hustota: 1,208 g/cm³ (25 °C) Bod vzplanutí: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274309 CAS číslo: 462-51-1 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...