3-Chloro-4-(trifluoromethoxy)benzoyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0179-10G
PC0179-5G
PC0179-1G
APOSPC0179-10GEA
7410
CZK
APOSPC0179-10G
APOSPC0179-5G
APOSPC0179-1G
3-Chloro-4-(trifluoromethoxy)benzoyl chloride 98%
3-Chloro-4-(trifluoromethoxy)benzoyl chloride
Vzorec:
C₈H₃Cl₂F₃O₂ MW: 259,009 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631558 CAS číslo: 261763-17-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...