Tisk…

3-Fluoro-2-iodobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Fluoro-2-iodophenyl)methanol

PC01679-1G PC01679-250MG PC01679-5G
APOSPC01679-1GEA 5230 CZK
APOSPC01679-1G APOSPC01679-250MG APOSPC01679-5G
3-Fluoro-2-iodobenzyl alcohol
3-Fluoro-2-iodobenzyl alcohol
Vzorec: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396946
CAS číslo: 1261827-82-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR