Tisk…

3-Iodo-1,1,1-trifluorohexane 95%

Dodavatel: Apollo Scientific
Varování

PC01659-1G PC01659-5G
APOSPC01659-1GEA 2570 CZK
APOSPC01659-1G APOSPC01659-5G
3-Iodo-1,1,1-trifluorohexane 95%
3-Iodo-1,1,1-trifluorohexane
Vzorec: C₆H₁₀F₃I
MW: 266.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606066
CAS číslo: 1257535-27-5
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR