2,5-Dichloro-3-fluorotoluene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,5-Dichloro-1-fluoro-3-methylbenzene
PC01531-5G
PC01531-1G
APOSPC01531-5GEA
5530
CZK
APOSPC01531-5G
APOSPC01531-1G
2,5-Dichloro-3-fluorotoluene 97%
2,5-Dichloro-3-fluorotoluene
Vzorec:
C₇H₅Cl₂F MW: 179,019 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16817664 CAS číslo: 1242339-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...