(2R)-1,1,1-Trifluoropropan-2-ol 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(R)-1,1,1-Trifluoropropan-2-ol
PC01525-1G
PC01525-250MG
PC01525-5G
APOSPC01525-1GEA
8270
CZK
APOSPC01525-1G
APOSPC01525-250MG
APOSPC01525-5G
(2R)-1,1,1-Trifluoropropan-2-ol 97%
(2R)-1,1,1-Trifluoropropan-2-ol
Vzorec:
C₃H₅F₃O Bod varu: 81…82 °C Hustota: 1,259 g/cm³ (25 °C) Bod vzplanutí: 18 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06797330 CAS číslo: 17628-73-8 UN: 1987 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...