Tisk…

5-Bromo-2-fluorothioanisole

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-1-fluoro-2-(methylsulphanyl)benzene, 5-Bromo-2-fluorophenyl methyl sulphide

PC01511-1G PC01511-5G
APOSPC01511-1GEA 4960 CZK
APOSPC01511-1G APOSPC01511-5G
5-Bromo-2-fluorothioanisole
5-Bromo-2-fluorothioanisole
Vzorec: C₇H₆BrFS
MW: 221.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056774
CAS číslo: 1370025-62-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR