Tisk…

3-Iodo-4-(trifluoromethoxy)nitrobenzene

Dodavatel: Apollo Scientific
Varování

PC01489-1G PC01489-5G PC01489-25G
APOSPC01489-1GEA 1990 CZK
APOSPC01489-1G APOSPC01489-5G APOSPC01489-25G
3-Iodo-4-(trifluoromethoxy)nitrobenzene
3-Iodo-4-(trifluoromethoxy)nitrobenzene
Vzorec: C₇H₃F₃INO₃
MW: 333,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194454
CAS číslo: 194344-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR