Tisk…

2-Fluoro-3-methylthiophenol 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-3-methylbenzenethiol, 2-Fluoro-3-methylphenyl mercaptan

PC01487-1G PC01487-250MG
APOSPC01487-1GEA 14220 CZK
APOSPC01487-1G APOSPC01487-250MG
2-Fluoro-3-methylthiophenol 96%
2-Fluoro-3-methylthiophenol
Vzorec: C₇H₇FS
MW: 142.194 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22481036
CAS číslo: 1243283-09-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR